missing translation for 'onlineSavingsMsg'
Learn More

1,1-Dimethyl-3-[3-(trifluoromethyl)phenyl]urea 97.0+%, TCI America™

Catalog No. D37735G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37735G 5 g
D377325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37735G Supplier TCI America Supplier No. D37735G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2164-17-2
Molecular Formula C10H11F3N2O
Molecular Weight (g/mol) 232.206
MDL Number MFCD00018012
InChI Key RZILCCPWPBTYDO-UHFFFAOYSA-N
Synonym fluometuron, 1,1-dimethyl-3-3-trifluoromethyl phenyl urea, higalcoton, pakhtaran, cotoran, lanex, cottonex, cotoran multi 50wp, herbicide c-2059, fluomethuron
PubChem CID 16562
ChEBI CHEBI:82012
IUPAC Name 1,1-dimethyl-3-[3-(trifluoromethyl)phenyl]urea
SMILES CN(C)C(=O)NC1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 162°C
Color White-Yellow
Quantity 5 g
Formula Weight 232.21
Percent Purity ≥97.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dimethyl-3-[3-(trifluoromethyl)phenyl]urea
RTECSNumber YT1575000
TSCA Yes