accessibility menu, dialog, popup

UserName

CAS RN 2164-17-2

IUPAC Name: 3,3-dimethyl-1-[3-(trifluoromethyl)phenyl]urea
Synonyms: fluometuron
Molecular Weight (g/mol): 232.21
Molecular Formula: C10H11F3N2O
InChi Key: RZILCCPWPBTYDO-UHFFFAOYSA-N
SMILES: CN(C)C(=O)NC1=CC=CC(=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results