Learn More
1,1-Cyclohexanediacetic acid, 98%
CAS: 4355-11-7 | C10H16O4 | 200.23 g/mol
$115.99 - $395.42
Chemical Identifiers
| CAS | 4355-11-7 |
|---|---|
| Molecular Formula | C10H16O4 |
| Molecular Weight (g/mol) | 200.23 |
| MDL Number | MFCD00001519 |
| InChI Key | YQPCHPBGAALCRT-UHFFFAOYSA-N |
| Synonym | 1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645 |
| PubChem CID | 78061 |
| IUPAC Name | 2-[1-(carboxymethyl)cyclohexyl]acetic acid |
| SMILES | C1CCC(CC1)(CC(=O)O)CC(=O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2081609
|
Thermo Scientific Chemicals
B2081609 |
10 g |
Each for $115.99
|
|
|||||
|
AAB2081618
|
Thermo Scientific Chemicals
B2081618 |
50 g |
Each for $395.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 4355-11-7 | |
| 200.23 | |
| YQPCHPBGAALCRT-UHFFFAOYSA-N | |
| 78061 | |
| C1CCC(CC1)(CC(=O)O)CC(=O)O |
| C10H16O4 | |
| MFCD00001519 | |
| 1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645 | |
| 2-[1-(carboxymethyl)cyclohexyl]acetic acid |
Specifications
| 4355-11-7 | |
| C10H16O4 | |
| 10 g | |
| YQPCHPBGAALCRT-UHFFFAOYSA-N | |
| 2-[1-(carboxymethyl)cyclohexyl]acetic acid | |
| 78061 | |
| 98% |
| 178°C to 182°C | |
| MFCD00001519 | |
| 1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645 | |
| C1CCC(CC1)(CC(=O)O)CC(=O)O | |
| 200.23 | |
| 200.23 | |
| 1,1-Cyclohexanediacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-427-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only