missing translation for 'onlineSavingsMsg'
Learn More

1,1-Cyclohexanediacetic acid, 98%

CAS: 4355-11-7 | C10H16O4 | 200.23 g/mol

Supplier:  Thermo Scientific Chemicals B2081618

 View more versions of this product

Catalog No. AAB2081618


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,1-Cyclohexanediacetic acid
178°C to 182°C
MFCD00001519
1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
C1CCC(CC1)(CC(=O)O)CC(=O)O
200.23
200.23
4355-11-7
C10H16O4
50 g
YQPCHPBGAALCRT-UHFFFAOYSA-N
2-[1-(carboxymethyl)cyclohexyl]acetic acid
78061
98%

Chemical Identifiers

4355-11-7
200.23
YQPCHPBGAALCRT-UHFFFAOYSA-N
78061
C1CCC(CC1)(CC(=O)O)CC(=O)O
C10H16O4
MFCD00001519
1,1-cyclohexanediacetic acid, 1,1-cyclohexanediaceticacid, cyclohexanediacetic acid, 2-1-carboxymethyl cyclohexyl acetic acid, 1,1-cyclohexane diacetic acid, 1-carboxymethyl cyclohexyl acetic acid, 2,2'-cyclohexane-1,1-diyl diacetic acid, 2-carboxymethyl cyclohexyl acetic acid, gabapentin impurity 2, pubchem13645
2-[1-(carboxymethyl)cyclohexyl]acetic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 224-427-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only