missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Bis(methylthio)-2-nitroethylene 98.0+%, TCI America™
Supplier: TCI America B1378100G
Specifications
| 1,1-Bis(methylthio)-2-nitroethylene | |
| 126°C | |
| MFCD00010443 | |
| 1,1-bis methylthio-2-nitroethylene, 1,1-bis methylsulfanyl-2-nitroethene, 1,1-dimethylthio-2-nitroethylene, 1-nitro-2,2-bis methylthio ethylene, ethene, 1,1-bis methylthio-2-nitro, 1,1-bis methylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethene, 1,1-dimethylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethylene | |
| CSC(=C[N+](=O)[O-])SC | |
| 165.225 | |
| 165.23 | |
| Crystalline Powder |
| 13623-94-4 | |
| C4H7NO2S2 | |
| 100 g | |
| NXGHEDHQXXXTTP-UHFFFAOYSA-N | |
| 1,1-bis(methylsulfanyl)-2-nitroethene | |
| 83623 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 13623-94-4 | |
| 165.225 | |
| NXGHEDHQXXXTTP-UHFFFAOYSA-N | |
| 83623 | |
| CSC(=C[N+](=O)[O-])SC |
| C4H7NO2S2 | |
| MFCD00010443 | |
| 1,1-bis methylthio-2-nitroethylene, 1,1-bis methylsulfanyl-2-nitroethene, 1,1-dimethylthio-2-nitroethylene, 1-nitro-2,2-bis methylthio ethylene, ethene, 1,1-bis methylthio-2-nitro, 1,1-bis methylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethene, 1,1-dimethylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethylene | |
| 1,1-bis(methylsulfanyl)-2-nitroethene |
Safety and Handling
TSCA : No