missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis(methylthio)-2-nitroethylene 98.0+%, TCI America™

Catalog No. p-7119023
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B137825G 25 g
B1378100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B137825G Supplier TCI America Supplier No. B137825G
Only null left

Chemical Identifiers

CAS 13623-94-4
Molecular Formula C4H7NO2S2
Molecular Weight (g/mol) 165.225
MDL Number MFCD00010443
InChI Key NXGHEDHQXXXTTP-UHFFFAOYSA-N
Synonym 1,1-bis methylthio-2-nitroethylene, 1,1-bis methylsulfanyl-2-nitroethene, 1,1-dimethylthio-2-nitroethylene, 1-nitro-2,2-bis methylthio ethylene, ethene, 1,1-bis methylthio-2-nitro, 1,1-bis methylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethene, 1,1-dimethylthio-2-nitroethene, 1,1-bis-methylthio-2-nitroethylene
PubChem CID 83623
IUPAC Name 1,1-bis(methylsulfanyl)-2-nitroethene
SMILES CSC(=C[N+](=O)[O-])SC

Specifications

Melting Point 126°C
Quantity 25 g
Formula Weight 165.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Bis(methylthio)-2-nitroethylene