missing translation for 'onlineSavingsMsg'
Learn More

1,1-Bis(4-hydroxyphenyl)cyclohexane 98.0+%, TCI America™

Catalog No. B1568500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1568500G 500 g
B156825G 25 g
B1568100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B1568500G Supplier TCI America Supplier No. B1568500G

Chemical Identifiers

CAS 843-55-0
Molecular Formula C18H20O2
Molecular Weight (g/mol) 268.356
MDL Number MFCD00019341
InChI Key SDDLEVPIDBLVHC-UHFFFAOYSA-N
PubChem CID 232446
IUPAC Name 4-[1-(4-hydroxyphenyl)cyclohexyl]phenol
SMILES C1CCC(CC1)(C2=CC=C(C=C2)O)C3=CC=C(C=C3)O

Specifications

Melting Point 189°C
Color White
Quantity 500 g
UN Number 3077
Synonym 4,4′C-Cyclohexylidenebisphenol
Formula Weight 268.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Bis(4-hydroxyphenyl)cyclohexane
EINECSNumber (4)-0044
TSCA No