missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Bis(4-hydroxyphenyl)cyclohexane 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 189°C |
| Color | White |
| Quantity | 25 g |
| UN Number | 3077 |
| Synonym | 4,4′C-Cyclohexylidenebisphenol |
| Formula Weight | 268.36 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,1-Bis(4-hydroxyphenyl)cyclohexane |
Safety and Handling
| EINECSNumber | (4)-0044 |
| TSCA | No |