missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-1,1'-Binaphthyl-2,2'-diyl Hydrogenphosphate 98.0+%, TCI America™
Supplier: TCI America B21305G
Specifications
| (+/-)-1,1′-Binaphthyl-2,2′-diyl Hydrogenphosphate | |
| C20H13O4P | |
| 5 g | |
| JEHUZVBIUCAMRZ-UHFFFAOYSA-N | |
| 348.294 | |
| 348.29 | |
| Crystalline Powder |
| 35193-63-6 | |
| MFCD00010045 | |
| 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide | |
| C1=CC=C2C(=C1)C=CC3=C2C4=C(C=CC5=CC=CC=C54)OP(=O)(O3)O | |
| 99589 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 35193-63-6 | |
| 348.294 | |
| JEHUZVBIUCAMRZ-UHFFFAOYSA-N | |
| 99589 |
| C20H13O4P | |
| MFCD00010045 | |
| 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide | |
| C1=CC=C2C(=C1)C=CC3=C2C4=C(C=CC5=CC=CC=C54)OP(=O)(O3)O |
Safety and Handling
TSCA : No