accessibility menu, dialog, popup

UserName

CAS RN 35193-63-6

IUPAC Name: 13-hydroxy-12,14-dioxa-13λ⁵-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-one
Synonyms: 13-hydroxy-12,14-dioxa-13λ⁵-phosphapentacyclo[13.8.0.0²,¹¹.0³,⁸.0¹⁸,²³]tricosa-1(23),2,4,6,8,10,15,17,19,21-decaen-13-one
Molecular Weight (g/mol): 348.29
Molecular Formula: C20H13O4P
InChi Key: JEHUZVBIUCAMRZ-UHFFFAOYSA-N
SMILES: OP1(=O)OC2=CC=C3C=CC=CC3=C2C2=C3C=CC=CC3=CC=C2O1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)