missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-1,1'-Binaphthyl-2,2'-diyl Hydrogenphosphate 98.0+%, TCI America™

Catalog No. p-7119500
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21305G 5 g
B213025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21305G Supplier TCI America Supplier No. B21305G
Only null left

Chemical Identifiers

CAS 35193-63-6
Molecular Formula C20H13O4P
Molecular Weight (g/mol) 348.294
MDL Number MFCD00010045
InChI Key JEHUZVBIUCAMRZ-UHFFFAOYSA-N
Synonym 1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, s-+-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, r---1,1'-binaphthyl-2,2'-diyl hydrogenphosphate, 1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r---bnp acid, 11bs-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, r---1,1'-binaphthyl-2,2'-diyl hydrogen phosphate, r-4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide, s-+-bnp acid, 4-hydroxydinaphtho 2,1-d:1',2'-f 1,3,2 dioxaphosphepine 4-oxide
PubChem CID 99589
SMILES C1=CC=C2C(=C1)C=CC3=C2C4=C(C=CC5=CC=CC=C54)OP(=O)(O3)O

Specifications

Quantity 5 g
Formula Weight 348.29
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (+/-)-1,1′-Binaphthyl-2,2′-diyl Hydrogenphosphate