missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate 98.0+%, TCI America™
Supplier: TCI America T345725G
Specifications
| 1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate | |
| Purple | |
| MFCD16621101 | |
| OBEJDMPKHNFDIA-UHFFFAOYSA-N | |
| 1,1,2,3-tetramethyl-1H-benzo[e]indol-3-ium; hexafluoro-λâµ-phosphanuide | |
| 11199491 | |
| ≥98.0% (HPLC,N) |
| 372081-65-7 | |
| C16H18F6NP | |
| 25 g | |
| F[P-](F)(F)(F)(F)F.CC1=[N+](C)C2=CC=C3C=CC=CC3=C2C1(C)C | |
| 369.29 | |
| 369.29 | |
| Crystalline Powder |
Chemical Identifiers
| 372081-65-7 | |
| 369.29 | |
| OBEJDMPKHNFDIA-UHFFFAOYSA-N | |
| 1,1,2,3-tetramethyl-1H-benzo[e]indol-3-ium; hexafluoro-λâµ-phosphanuide |
| C16H18F6NP | |
| MFCD16621101 | |
| 11199491 | |
| F[P-](F)(F)(F)(F)F.CC1=[N+](C)C2=CC=C3C=CC=CC3=C2C1(C)C |
Safety and Handling
TSCA : No