missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate 98.0+%, TCI America™

$175.22 - $518.21

Chemical Identifiers

CAS 372081-65-7
Molecular Formula C16H18F6NP
Molecular Weight (g/mol) 369.29
MDL Number MFCD16621101
InChI Key OBEJDMPKHNFDIA-UHFFFAOYSA-N
PubChem CID 11199491
IUPAC Name 1,1,2,3-tetramethyl-1H-benzo[e]indol-3-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CC1=[N+](C)C2=CC=C3C=CC=CC3=C2C1(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
T34575G
View Documents
TCI America
T34575G
5 g N/A
Only null left
 
T345725G
View Documents
TCI America
T345725G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

372081-65-7
369.29
OBEJDMPKHNFDIA-UHFFFAOYSA-N
1,1,2,3-tetramethyl-1H-benzo[e]indol-3-ium; hexafluoro-λ⁵-phosphanuide
C16H18F6NP
MFCD16621101
11199491
F[P-](F)(F)(F)(F)F.CC1=[N+](C)C2=CC=C3C=CC=CC3=C2C1(C)C

Specifications

372081-65-7
C16H18F6NP
5 g
F[P-](F)(F)(F)(F)F.CC1=[N+](C)C2=CC=C3C=CC=CC3=C2C1(C)C
369.29
369.29
Crystalline Powder
Purple
MFCD16621101
OBEJDMPKHNFDIA-UHFFFAOYSA-N
1,1,2,3-tetramethyl-1H-benzo[e]indol-3-ium; hexafluoro-λ⁵-phosphanuide
11199491
≥98.0% (HPLC,N)
1,1,2,3-Tetramethyl-1H-benzo[e]indolium Hexafluorophosphate
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'