accessibility menu, dialog, popup

UserName

CH3NH3CCH3CH3FPFFFFF

CAS RN 372081-65-7

IUPAC Name: 1,1,2,3-tetramethyl-1H-benzo[e]indol-3-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 1,1,2,3-tetramethylbenzo[e]indol-3-ium hexafluorophosphate
Molecular Weight (g/mol): 369.29
Molecular Formula: C16H18F6NP
InChi Key: OBEJDMPKHNFDIA-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CC1=[N+](C)C2=CC=C3C=CC=CC3=C2C1(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results