missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrabromoethane 98.0+%, TCI America™

Catalog No. T003425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T003425G 25 g
T0034500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T003425G Supplier TCI America Supplier No. T003425G

Chemical Identifiers

CAS 79-27-6
Molecular Formula C2H2Br4
Molecular Weight (g/mol) 345.65
MDL Number MFCD00000133
InChI Key QXSZNDIIPUOQMB-UHFFFAOYSA-N
Synonym acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o
PubChem CID 6588
IUPAC Name 1,1,2,2-tetrabromoethane
SMILES BrC(Br)C(Br)Br

Specifications

Melting Point 0°C
Color Yellow
Boiling Point 243°C
Quantity 25 g
UN Number 2504
Formula Weight 345.65
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,2,2-Tetrabromoethane
EINECSNumber (2)-0077
RTECSNumber KI8225000
TSCA Yes