accessibility menu, dialog, popup

UserName

BrBrBrBr

CAS RN 79-27-6

IUPAC Name: 1,1,2,2-tetrabromoethane
Synonyms: tetrabromoethane
Molecular Weight (g/mol): 345.65
Molecular Formula: C2H2Br4
InChi Key: QXSZNDIIPUOQMB-UHFFFAOYSA-N
SMILES: BrC(Br)C(Br)Br
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (2)
  • (2)
  • (2)
  • (6)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (5)
  • (2)