missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrabromoethane, Practical, 95%, Spectrum™ Chemical

Catalog No. p-7166626
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly-Coated Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608016 500 g Amber Glass Bottle
18608017 5 kg Poly-Coated Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18608016 Supplier Spectrum Chemical Mfg Cor Supplier No. T1022500GM
Only null left

Chemical Identifiers

CAS 79-27-6

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 0.95
Grade Practical

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.