missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrabromoethane, 98%

CAS: 79-27-6 | C2H2Br4 | 345.65 g/mol

Supplier:  Thermo Scientific Chemicals 180870025

 View more versions of this product

Catalog No. AC180870025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1,1,2,2-Tetrabromoethane
97.5
1.0°C
244.0°C
97.5% min. (GC)
C2H2Br4
Br2CHCHBr2
2.5 kg
2.96
acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o
QXSZNDIIPUOQMB-UHFFFAOYSA-N
1,1,2,2-tetrabromoethane
6588
98%
79-27-6
100.0
2.9600g/mL
Authentic
Glass bottle
1.6355 to 1.6380
MFCD00000133
01, 94
15, 9330
Solubility in water: 0.63g/L (20°C). Other solubilities: soluble in acetone, benzene, miscible with ethanol, ether, chloroform, aniline, and glacial acetic acid
BrC(Br)C(Br)Br
345.65
345.64
Liquid

Chemical Identifiers

79-27-6
345.65
QXSZNDIIPUOQMB-UHFFFAOYSA-N
6588
BrC(Br)C(Br)Br
C2H2Br4
MFCD00000133
acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o
1,1,2,2-tetrabromoethane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Fatal if inhaled.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 201-191-5

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only