missing translation for 'onlineSavingsMsg'
Learn More

1,1,2,2-Tetrabromoethane, 97%

Catalog No. AAA12943AD
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA12943AD 50 mL
AAA12943AK 250 mL
AAA12943AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA12943AD Supplier Thermo Scientific Chemicals Supplier No. A12943AD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 79-27-6 | C2H2Br4 | 345.65 g/mol

1,1,2,2-Tetrabromoethane is useful for the separation of minerals from supporting rock. It is used as a solvent for fats, greases, oils and waxes. It acts as a mercury substitute in the production of gauges and balancing equipment. It finds application as a floating agent used in processing and separation of mineral oils. Further, it is used as refractive index liquid in microscopy. In addition to this, it is also employed as a flame-retardant additive for polystyrenes, polyurethanes and polyolefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,2,2-Tetrabromoethane is useful for the separation of minerals from supporting rock. It is used as a solvent for fats, greases, oils and waxes. It acts as a mercury substitute in the production of gauges and balancing equipment. It finds application as a floating agent used in processing and separation of mineral oils. Further, it is used as refractive index liquid in microscopy. In addition to this, it is also employed as a flame-retardant additive for polystyrenes, polyurethanes and polyolefins.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, aluminum, magnesium, alkali metals and zinc.

Chemical Identifiers

CAS 79-27-6
Molecular Formula C2H2Br4
Molecular Weight (g/mol) 345.65
MDL Number MFCD00000133
InChI Key QXSZNDIIPUOQMB-UHFFFAOYSA-N
Synonym acetylene tetrabromide, muthmann's liquid, ethane, 1,1,2,2-tetrabromo, tetrabromoacetylene, sym-tetrabromoethane, s-tetrabromoethane, 1,1,2,2-tetrabromaethan, 1,1,2,2-tetrabromoetano, tbe, unii-v00gkc584o
PubChem CID 6588
IUPAC Name 1,1,2,2-tetrabromoethane
SMILES BrC(Br)C(Br)Br

Specifications

Density 2.967
Boiling Point 118°C to 120°C (15 mmHg)
Refractive Index 1.637
Quantity 50 mL
UN Number UN2504
Merck Index 14,9185
Solubility Information Immiscible with water.
Formula Weight 345.67
Percent Purity 97%
Chemical Name or Material 1,1,2,2-Tetrabromoethane
Hazard Category H319-H330
Hazard Statement GHS H Statement
H330-H319
Fatal if inhaled.
Causes serious eye irritation.
Precautionary Statement P260-P271-P284-P304+P340-P305+P351+P338-P310-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TETRABROMOETHANE
EINECSNumber 201-191-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only