missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1,1',1'-Tetramethyl-4,4'-(methylenedi-p-phenylene)disemicarbazide 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 177°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 370.46 |
| Percent Purity | ≥95.0% (HPLC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 1,1,1′,1′-Tetramethyl-4,4′-(methylenedi-p-phenylene)disemicarbazide |
Safety and Handling
| EINECSNumber | (4)-1450 |
| TSCA | No |