CAS RN 85095-61-0
CAS RN 85095-61-0
IUPAC Name:
3-(dimethylamino)-1-[4-({4-[(N',N'-dimethylhydrazinecarbonyl)amino]phenyl}methyl)phenyl]urea
Synonyms:
3-(dimethylamino)-1-[4-({4-[(N',N'-dimethylhydrazinecarbonyl)amino]phenyl}methyl)phenyl]urea
Molecular Weight (g/mol):
370.46
Molecular Formula:
C19H26N6O2
InChi Key:
AQABZFKTYXFIJF-UHFFFAOYSA-N
SMILES:
CN(C)NC(=O)NC1=CC=C(CC2=CC=C(NC(=O)NN(C)C)C=C2)C=C1
1
–
1
of
1
results
1
1,1,1',1'-Tetramethyl-4,4'-(methylenedi-p-phenylene)disemicarbazide 95.0+%, TCI America™
CAS: 85095-61-0 Molecular Formula: C19H26N6O2 Molecular Weight (g/mol): 370.457 MDL Number: MFCD00525631 InChI Key: AQABZFKTYXFIJF-UHFFFAOYSA-N Synonym: Bis[4-[3-(dimethylamino)ureido]phenyl]methane PubChem CID: 12842685 IUPAC Name: 1-(dimethylamino)-3-[4-[[4-(dimethylaminocarbamoylamino)phenyl]methyl]phenyl]urea SMILES: CN(C)NC(=O)NC1=CC=C(C=C1)CC2=CC=C(C=C2)NC(=O)NN(C)C