missing translation for 'onlineSavingsMsg'
Learn More

1,1,1',1'-Tetramethyl-4,4'-(methylenedi-p-phenylene)disemicarbazide 95.0+%, TCI America™

Catalog No. T205125G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T205125G 25 g
T2051500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T205125G Supplier TCI America Supplier No. T205125G
Only null left

Chemical Identifiers

CAS 85095-61-0
Molecular Formula C19H26N6O2
Molecular Weight (g/mol) 370.457
MDL Number MFCD00525631
InChI Key AQABZFKTYXFIJF-UHFFFAOYSA-N
Synonym Bis[4-[3-(dimethylamino)ureido]phenyl]methane
PubChem CID 12842685
IUPAC Name 1-(dimethylamino)-3-[4-[[4-(dimethylaminocarbamoylamino)phenyl]methyl]phenyl]urea
SMILES CN(C)NC(=O)NC1=CC=C(C=C1)CC2=CC=C(C=C2)NC(=O)NN(C)C

Specifications

Melting Point 177°C
Color White
Quantity 25 g
Formula Weight 370.46
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1,1′,1′-Tetramethyl-4,4′-(methylenedi-p-phenylene)disemicarbazide
EINECSNumber (4)-1450
TSCA No