missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EDTA (0.5 M), pH 8.0

Catalog No. FERR1021
Change view
Click to view available options
Quantity:
5 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR1021 5 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR1021 Supplier Thermo Scientific™ Supplier No. R1021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimize biochemistry or molecular biology applications that require a chelator for divalent cations.

0.5 M EDTA, pH 8.0 is an aqueous solution of 0.22 μm membrane-filtered ethylendiaminetetraacetic acid (EDTA), sodium salt. It's ideal for biochemistry or molecular biology applications that require a chelator for divalent cations. By forming complexes with metal ions, EDTA inhibits metal-dependent enzymatic reactions. EDTA complexes metal ions at a 1:1 molar ratio.

Nucleic acids are normally stored in TE buffer that contains 1 mM EDTA, which prevents degradation of nucleic acids by metal dependent nucleases.

Applications
  • Termination of metal-dependent enzymatic reactions
  • Protection of DNA against metal-dependent nucleases

Specifications

Chemical Name or Material EDTA
Product Type EDTA
Quantity 5 x 1 mL
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.