missing translation for 'onlineSavingsMsg'
Learn More

0.07% n-Hexylamine in 60/40 Acetonitrile, HPLC, Reagents

Catalog No. 01334698
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334698 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01334698 Supplier Labchem Inc Supplier No. CN0003004C

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

0.07% n-Hexylamine in 60/40 Acetonitrile, HPLC, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 75-05-8 , 7732-18-5 , 64-19-7
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym n-Hexylamine, 1-Aminohexane
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Color Colorless
Boiling Point 76°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.8741
Percent Purity 60%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 0.07% n-Hexylamine in 60/40 Acetonitrile, HPLC