Get an Accessory with Labconco Freeze Dryer Purchase
When you purchase a qualifying Labconco™ FreeZone™ Benchtop or Console Freeze Dryer, you’ll get your choice of either an 8-port manifold or 12-port drying chamber at no additional cost.
Once you’ve made your qualifying purchase, log in to the Redemption Center to redeem this offer.
Features and Benefits
Labconco FreeZone Freeze Dryers are designed to deliver consistent, high-quality results to make lyophilizing challenging samples easy. FreeZone Freeze Dryers include a large, touchscreen display, Lyo-Worksâ„¢ Operating System for on-board controls, program development, and data recording to easily monitor process parameters.
Labconco™ FreeZone™ 2.5 L -50°C Benchtop Freeze Dryer, 115 V US Model
FreeZone Benchtop Freeze Dryers are designed for lyophilizing light to moderate sample loads. The compact design is ideal for labs with limited floor space. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Shipping Weight (Metric) | 48.5 kg |
---|---|
Height (Metric) | 42.9 cm |
Depth (English) | 19.2 in. |
Plug Type | North America |
Width (English) | 14.5 in. |
Capacity (Metric) | 2.5 L (before defrosting) |
Material | Steel |
Width (Metric) | 36.8 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 48.8 cm |
Voltage | 115 V |
Amperage | 14 A |
Frequency | 60 Hz |
Temperature Range (English) | -58°F |
Certifications/Compliance | UL, CAN/CSA, CE |
Temperature Range (Metric) | -50°C |
Height (English) | 16.9 in. |
Labconco™ FreeZone™ 4.5 L -50°C Benchtop Freeze Dryer, 115 V US Model
FreeZone Benchtop Freeze Dryers are designed for lyophilizing light to moderate sample loads. The compact design is ideal for labs with limited floor space. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Shipping Weight (Metric) | 49.4 kg |
---|---|
Height (Metric) | 48 cm |
Depth (English) | 22.4 in. |
Plug Type | North America |
Width (English) | 16.5 in. |
Capacity (Metric) | 4.5 L (before defrosting) |
Material | Steel |
Width (Metric) | 41.9 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 56.8 cm |
Voltage | 115 V |
Amperage | 14 A |
Frequency | 60 Hz |
Temperature Range (English) | -58°F |
Certifications/Compliance | UL, CAN/CSA, CE |
Temperature Range (Metric) | -50°C |
Height (English) | 18.9 in. |
Labconco™ FreeZone™ 4.5 L -84°C Benchtop Freeze Dryer, 230 V Model
FreeZone 4.5 Liter Benchtop Freeze Dryers are designed for lyophilizing light to moderate sample loads. The compact design is ideal for labs with limited floor space. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Shipping Weight (Metric) | 75.3 kg |
---|---|
Height (Metric) | 46.9 cm |
Depth (English) | 26.5 in. |
Width (English) | 22.5 in. |
Capacity (Metric) | 2.5 L (before defrosting) |
Material | Steel |
Width (Metric) | 57.1 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 67.3 cm |
Voltage | 230 V |
Amperage | 10 A |
Frequency | 60 Hz |
Temperature Range (English) | -119°C |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 166 lb. |
Temperature Range (Metric) | -84°C |
Height (English) | 18.5 in. |
Labconco™ FreeZone™ 2.5 L -50°C Benchtop Freeze Dryer, 230 V Model
FreeZone 2.5 L Benchtop Freeze Dryer designed for lyophilizing light sample loads. Compact design ideal for labs with limited floor space. Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display. Protect samples with remote notification and data recording.
Shipping Weight (Metric) | 48.53 kg |
---|---|
Height (Metric) | 42.92 cm |
Depth (English) | 19.2 in. |
Width (English) | 14.5 in. |
Capacity (Metric) | 2.5 L (before defrosting) |
Material | Steel |
Width (Metric) | 36.83 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 48.76 cm |
Voltage | 230 V |
Amperage | 7 A |
Frequency | 60 Hz |
Temperature Range (English) | -58°F |
Certifications/Compliance | UL, CAN/CSA, CE |
Temperature Range (Metric) | -50°C |
Height (English) | 16.9 in. |
Labconco™ FreeZone™ 4.5 L -105°C Benchtop Freeze Dryer, 115 V US Model
FreeZone Benchtop Freeze Dryers are designed for lyophilizing light to moderate sample loads. The compact design is ideal for labs with limited floor space. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Shipping Weight (Metric) | 78 kg |
---|---|
Height (Metric) | 46.9 cm |
Depth (English) | 26.5 in. |
Plug Type | North America |
Width (English) | 22.5 in. |
Capacity (Metric) | 4.5 L (before defrosting) |
Material | Steel |
Width (Metric) | 57.1 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 67.3 cm |
Voltage | 115 V |
Amperage | 16 A |
Frequency | 60 Hz |
Temperature Range (English) | -157°F |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 172 lb. |
Temperature Range (Metric) | -105°C |
Height (English) | 18.5 in. |
Labconco™ FreeZone™ 8L -50°C Benchtop Freeze Dryers, 230V Models
FreeZone 8L Benchtop Freeze Dryers are designed for lyophilizing moderate sample loads. The compact design is ideal for labs with limited floor space. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Shipping Weight (Metric) | 59 kg |
---|---|
Height (Metric) | 46.9 cm |
Depth (English) | 26.5 in. |
Width (English) | 22.5 in. |
Capacity (Metric) | 8 L (before defrosting) |
Material | Steel |
Width (Metric) | 57.1 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 67.3 cm |
Voltage | 230 V |
Amperage | 7A |
Frequency | 60 Hz |
Temperature Range (English) | -58°F |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 130 lb. |
Temperature Range (Metric) | -50°C |
Height (English) | 18.5 in. |
Labconco™ FreeZone™ 6L -50°C Console Freeze Dryers, 115V US Models
FreeZone 6 Liter Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Shipping Weight (Metric) | 115.7 kg |
---|---|
Height (Metric) | 92.9 cm |
Depth (English) | 29.6 in. |
Plug Type | North America |
Width (English) | 31.8 in. |
Capacity (Metric) | 6 L (before defrosting) |
Material | Steel |
Width (Metric) | 80.7 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 75.1 cm |
Voltage | 115 V |
Amperage | 20A |
Frequency | 60 Hz |
Temperature Range (English) | -58°F |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 255 lb. |
Temperature Range (Metric) | -50°C |
Height (English) | 36.6 in. |
Labconco™ FreeZone™ 6L -84°C Console Freeze Dryers, 230V Models
FreeZone 6L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Worksâ„¢ OS provides flexibility and convenience in easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
Shipping Weight (Metric) | 122.5 kg |
---|---|
Height (Metric) | 92.9 cm |
Depth (English) | 29.6 in. |
Width (English) | 31.8 in. |
Capacity (Metric) | 6 L (before defrosting) |
Material | Steel |
Width (Metric) | 80.7 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 75.1 cm |
Voltage | 230 V |
Amperage | 10 A |
Temperature Range (English) | -119°C |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 270 lb. |
Temperature Range (Metric) | -84°C |
Height (English) | 36.6 in. |
Labconco™ FreeZone™ 12L -50°C Console Freeze Dryers
FreeZone 12L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Worksâ„¢ OS provides flexibility and convenience in easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
Shipping Weight (Metric) | 124.7 kg |
---|---|
Height (Metric) | 92.9 cm |
Depth (English) | 29.6 in. |
Width (English) | 31.8 in. |
Capacity (Metric) | 12 L (before defrosting) |
Material | Steel |
Width (Metric) | 80.7 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 75.1 cm |
Voltage | 230 V |
Amperage | 9A |
Temperature Range (English) | -58°F |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 275 lb. |
Temperature Range (Metric) | -50°C |
Height (English) | 36.6 in. |
Labconco™ FreeZone™ 12L -84°C Console Freeze Dryers, Purge Valve and Mini Chamber
FreeZoneâ„¢ 12L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
Shipping Weight (Metric) | 151.95 kg |
---|---|
Height (Metric) | 92.9 cm |
Depth (English) | 29.6 in. |
Width (English) | 31.8 in. |
Capacity (Metric) | 12 L (before defrosting) |
Material | Steel |
Width (Metric) | 80.7 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 75.1 cm |
Voltage | 230 V |
Frequency | 60 Hz |
Temperature Range (English) | -119°C |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 335 lb. |
Temperature Range (Metric) | -84°C |
Height (English) | 36.6 in. |
Labconco™ FreeZone™ 6L -50°C Console Freeze Dryers, Stoppering Tray Dryer, 230V Models
FreeZoneâ„¢ 6L Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials under vacuum. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
Shipping Weight (Metric) | 307.53 kg |
---|---|
Height (Metric) | 161.7 cm |
Depth (English) | 29.6 in. |
Width (English) | 82.2 in. |
Capacity (Metric) | 6 L (before defrosting) |
Material | Steel |
Width (English) Inside | 32.4 in. |
Type | Freeze Dryer/Lyophilizer |
Depth (Metric) | 75.1 cm |
Voltage | 230 V |
Frequency | 60 Hz |
Temperature Range (English) | -58°F |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 678 lb. |
Temperature Range (Metric) | -50°C |
Height (English) | 63.7 in. |
Labconco™ FreeZone™ 6L -50°C Console Freeze Dryers, Purge Valve and Mini Chamber, 230V Models
FreeZone 6L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Worksâ„¢ OS provides flexibility and convenience in easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
Shipping Weight (Metric) | 124.73 kg |
---|---|
Height (Metric) | 92.9 cm |
Depth (English) | 29.6 in. |
Width (English) | 31.8 in. |
Capacity (Metric) | 6 L (before defrosting) |
Material | Steel |
Width (Metric) | 80.7 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 75.1 cm |
Voltage | 230 V |
Amperage | 8A |
Temperature Range (English) | -58°F |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 275 lb. |
Temperature Range (Metric) | -50°C |
Height (English) | 36.6 in. |
Labconco™ FreeZone™ 6L Console Freeze Dryers, -84°C, 115V US Models
FreeZone 6 Liter Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display.
Shipping Weight (Metric) | 122.5 kg |
---|---|
Height (Metric) | 92.9 cm |
Depth (English) | 29.6 in. |
Plug Type | North America |
Width (English) | 31.8 in. |
Capacity (Metric) | 6 L (before defrosting) |
Material | Steel |
Width (Metric) | 80.7 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 0 |
Depth (Metric) | 75.1 cm |
Voltage | 115 V |
Amperage | 16A |
Frequency | 60 Hz |
Temperature Range (English) | -119°C |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 270 lb. |
Temperature Range (Metric) | -84°C |
Height (English) | 36.6 in. |
Labconco™ FreeZone™ 18L -50°C Console Freeze Dryers
FreeZoneâ„¢ 18L Console Freeze Dryers are designed for lyophilizing moderate to large sample loads or numerous small container batches. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
Width (English) | 31.8 in. |
---|---|
No. of Shelves | 0 |
Voltage | 230 V |
Temperature Range (Metric) | -50°C |
Labconco™ FreeZone™ 18L -50°C Console Freeze Dryers w/Stoppering Tray Dryer, 230V Models
FreeZone 18L Console Freeze Dryers with Stoppering Tray Dryers are designed for lyophilizing moderate to large loads and stoppering vials or serum bottles under vacuum. New Lyo-Worksâ„¢ OS provides flexibility and convenience in an easy-to-use, touch screen display. Your samples are protected with remote notification and data recording.
Shipping Weight (Metric) | 323.41 kg |
---|---|
Height (Metric) | 161.7 cm |
Depth (English) | 29.6 in. |
Width (English) | 32.4 in. |
Capacity (Metric) | 18 L (before defrosting) |
Material | Steel |
Width (Metric) | 82.2 cm |
Type | Freeze Dryer/Lyophilizer |
No. of Shelves | 3 |
Depth (Metric) | 75.1 cm |
Voltage | 230 V |
Temperature Range (English) | -58°C |
Certifications/Compliance | UL, CAN/CSA, CE |
Shipping Weight (English) | 713 lb. |
Temperature Range (Metric) | -50°C |
Height (English) | 63.7 in. |
Catalog Number | Description | |||
---|---|---|---|---|
Catalog Number |
Description Labconco 8-Port Manifold for FreeZone Freeze Dryers | Learn More | ||
Catalog Number |
Description Labconco Drying Chamber for FreeZone Freeze-Dry Systems, 12-Port | Learn More |