UserName
Fisherbrand Microscopy Solutions
Outstanding Performance and Versatility
Microscopes and accessories for any application.
Upright, Stereo, and Inverted Microscopes
Fisherbrand Entry Level Research Grade Inverted Microscope
Perfect for classroom observations of living cells and organisms under more natural conditions.
  • Siedentopf head design for more flexible eyepiece adjustment
  • Infinity-corrected optics for consistency between objectives
  • Extra-long working distance (up to 195mm)
  • Koehler LED illumination for uniform and glare-free viewing
Shop Now
Inverted Microscopes
Upright Compound Microscopes
Fisherbrand Research Grade Upright Microscopes are available in a number of models and with a range of features.
Choose advanced models for photography and more demanding research, research models for most clinical and research lab use, or entry-level models for general laboratory demands.
Stereo and Stereo Zoom Microscopes
Zoom stereo microscopes are available in research grade or entry level, with corresponding features and options. Or choose an entry-level stereo microscope with dual, fixed magnification for more routine use.
Microscope Accessories
Customize your microscopes to modify magnification or replace damaged or broken parts.
Filename
fisherbrand-entry-level-inverted-microscope-data-sheet.pdf
Size
725 KB
Format
application/pdf
Filename
fisherbrand-advanced-upright-microscope-data-sheet.pdf
Size
530 KB
Format
application/pdf
Filename
fisherbrand-research-grade-upright-microscope-data-sheet.pdf
Size
651 KB
Format
application/pdf
Learn about the features that make this microscope perfect for both educational purposes and basic lab use.
Filename
fisherbrand-entry-level-upright-microscope-data-sheet.pdf
Size
448 KB
Format
application/pdf
Filename
fisherbrand-research-grade-stereo-zoom-microscope-data-sheet.pdf
Size
228 KB
Format
application/pdf
Filename
fisherbrand-entry-level-stereo-zoom-microscopes-data-sheet.pdf
Size
272 KB
Format
application/pdf
Learn about the unique features that make this microscope ideal for high school classrooms.
Filename
fisherbrand-research-grade-stereo-microscopes-data-sheet.pdf
Size
236 KB
Format
application/pdf
Find out more about this basic, fixed magnification stereo microscope, which includes features that make it perfect for simple applications.
Filename
fisherbrand-entry-level-stereo-microscopes-data-sheet.pdf
Size
273 KB
Format
application/pdf