accessibility menu, dialog, popup

UserName

CAS RN 99071-55-3

IUPAC Name: 2-[4-(cyanomethyl)phenyl]acetamide
Synonyms: 2-[4-(cyanomethyl)phenyl]acetamide
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: QNLWFGMDHQLLHX-UHFFFAOYSA-N
SMILES: NC(=O)CC1=CC=C(CC#N)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results