accessibility menu, dialog, popup

UserName

(R)(R)(R)(R)(S)(S)OOO(S)(S)(R)(R)HOOHOHHOHOOHHOOH

CAS RN 99-20-7

IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol
Synonyms: α,α'-trehalose
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: HDTRYLNUVZCQOY-LBYRQFQKNA-N
SMILES: OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
Physical Form 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (6)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)