missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Trehalose, anhydrous

Catalog No. AAJ6600618
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6600618 50 g
AAJ6600606 5 g
AAJ6600609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6600618 Supplier Thermo Scientific Chemicals Supplier No. J6600618

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-20-7 | C12H22O11 | 342.30 g/mol

Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Trehalose, Dihydrate, is an inducer of autophagy, it protect cells against numerous environmental stresses. It is seldom used as a direct replacement for conventional sweeteners. Used as a cryoprotectant in a variety of cell freezing media.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 99-20-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006628
InChI Key HDTRYLNUVZCQOY-LBYRQFQKNA-N
Synonym trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose
PubChem CID 7427
ChEBI CHEBI:16551
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Color White
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Optical Rotation +197° (c=7 in Water)
Formula Weight 342.3
Physical Form Powder
Chemical Name or Material D-(+)-Trehalose, anhydrous
EINECSNumber 202-739-6
RTECSNumber LZ5776770
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only