missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Trehalose Anhydrous 98.0+%, TCI America™

Catalog No. T08325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T08325G 5 g
T083225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T08325G Supplier TCI America Supplier No. T08325G

missing translation for 'validMainframeMsg'

CAS: 99-20-7 | C12H22O11 | 342.30 g/mol

Chemical Identifiers

CAS 99-20-7
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00006628
InChI Key HDTRYLNUVZCQOY-LBYRQFQKNA-N
Synonym trehalose, d-trehalose, alpha,alpha-trehalose, mycose, ergot sugar, alpha-d-trehalose, d-+-trehalose, d-+-trehalose, anhydrous, alpha,alpha'-trehalose, trehaose
PubChem CID 7427
ChEBI CHEBI:16551
IUPAC Name (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-{[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}oxane-3,4,5-triol
SMILES OC[C@H]1O[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 205°C
Color White
Quantity 5 g
Formula Weight 342.30
Percent Purity ≥98.0% (GC)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material D-(+)-Trehalose Anhydrous
EINECSNumber (8)-0674
TSCA Yes