accessibility menu, dialog, popup

UserName

CAS RN 99-03-6

IUPAC Name: 1-(3-aminophenyl)ethan-1-one
Synonyms: M-aminoacetophenone
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: CKQHAYFOPRIUOM-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC(N)=C1
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)