missing translation for 'onlineSavingsMsg'
Learn More

3'-Aminoacetophenone 98.0+%, TCI America™

Catalog No. A024925G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A024925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A024925G Supplier TCI America Supplier No. A024925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 99-03-6
Melting Point 98°C
Boiling Point 252°C
Molecular Formula C8H9NO
Quantity 25 g
MDL Number MFCD00007796
Synonym 3'-aminoacetophenone, 1-3-aminophenyl ethanone, m-aminoacetophenone, 3-aminoacetophenone, 3-acetylaniline, m-acetylaniline, ethanone, 1-3-aminophenyl, 1-3-aminophenyl ethan-1-one, m-aminoacetylbenzene, acetophenone, 3'-amino
InChI Key CKQHAYFOPRIUOM-UHFFFAOYSA-N
SMILES CC(=O)C1=CC=CC(N)=C1
IUPAC Name 1-(3-aminophenyl)ethan-1-one
Molecular Weight (g/mol) 135.17
PubChem CID 7417
Formula Weight 135.17
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Aminoacetophenone
Show More Show Less
EINECSNumber (3)-1242
RTECSNumber AM5800000
TSCA Yes