missing translation for 'onlineSavingsMsg'
Learn More

3'-Aminoacetophenone, 97%

Catalog No. AAA1250330
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1250330 250 g
AAA1250318 50 g
AAA125030B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1250330 Supplier Thermo Scientific Chemicals Supplier No. A1250330
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 99-03-6 | C8H9NO | 135.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99-03-6
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.17
MDL Number MFCD00007796
InChI Key CKQHAYFOPRIUOM-UHFFFAOYSA-N
Synonym 3'-aminoacetophenone, 1-3-aminophenyl ethanone, m-aminoacetophenone, 3-aminoacetophenone, 3-acetylaniline, m-acetylaniline, ethanone, 1-3-aminophenyl, 1-3-aminophenyl ethan-1-one, m-aminoacetylbenzene, acetophenone, 3'-amino
PubChem CID 7417
IUPAC Name 1-(3-aminophenyl)ethanone
SMILES CC(=O)C1=CC=CC(N)=C1

Specifications

Melting Point 94°C to 98°C
Boiling Point 289°C to 291°C
Quantity 250 g
Beilstein 386009
Merck Index 14,413
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 3'-Aminoacetophenone
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-722-3
RTECSNumber AM5800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only