CAS RN 98919-68-7
IUPAC Name:
2-phenylcyclohexan-1-ol
Synonyms:
cyclohexanol, 2-phenyl-
Molecular Weight (g/mol):
176.26
Molecular Formula:
C12H16O
InChi Key:
AAIBYZBZXNWTPP-UHFFFAOYNA-N
SMILES:
OC1CCCCC1C1=CC=CC=C1
1
–
1
of
1
results
1
(1R,2S)-(-)-trans-2-Phenyl-1-cyclohexanol 98.0+%, TCI America™
CAS: 98919-68-7 Molecular Formula: C12H16O Molecular Weight (g/mol): 176.259 MDL Number: MFCD00075488 InChI Key: AAIBYZBZXNWTPP-NWDGAFQWSA-N PubChem CID: 102793 IUPAC Name: (1R,2S)-2-phenylcyclohexan-1-ol SMILES: C1CCC(C(C1)C2=CC=CC=C2)O