missing translation for 'onlineSavingsMsg'
Learn More

(1R,2S)-(-)-trans-2-Phenyl-1-cyclohexanol 98.0+%, TCI America™

Catalog No. T14901G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T14901G 1 g
T1490100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T14901G Supplier TCI America Supplier No. T14901G
Only null left

Chemical Identifiers

CAS 98919-68-7
Molecular Formula C12H16O
Molecular Weight (g/mol) 176.259
MDL Number MFCD00075488
InChI Key AAIBYZBZXNWTPP-NWDGAFQWSA-N
PubChem CID 102793
IUPAC Name (1R,2S)-2-phenylcyclohexan-1-ol
SMILES C1CCC(C(C1)C2=CC=CC=C2)O

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 105°C
Quantity 1 g
Formula Weight 176.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (1R,2S)-(-)-trans-2-Phenyl-1-cyclohexanol