accessibility menu, dialog, popup

UserName

CH3NNH3CH3C

CAS RN 98892-75-2

IUPAC Name: 3-butyl-1,2-dimethyl-1H-imidazol-3-ium
Synonyms: 1-butyl-2,3-dimethylimidazol-1-ium
Molecular Weight (g/mol): 153.25
Molecular Formula: C9H17N2
InChi Key: XUAXVBUVQVRIIQ-UHFFFAOYSA-N
SMILES: CCCC[N+]1=C(C)N(C)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)