missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-2,3-dimethylimidazolium chloride, 99%

Catalog No. AAH2727018
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2727018 50 g
AAH2727006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2727018 Supplier Thermo Scientific Chemicals Supplier No. H2727018

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98892-75-2 | C9H17ClN2 | 188.699 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98892-75-2
Molecular Formula C9H17ClN2
Molecular Weight (g/mol) 188.699
MDL Number MFCD03093291
InChI Key HHHYPTORQNESCU-UHFFFAOYSA-M
Synonym 1-butyl-2,3-dimethylimidazolium chloride, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium chloride, 1-butyl-2,3-dimethylimidazol-1-ium chloride, 3-butyl-1,2-dimethylimidazol-1-ium chloride, acmc-209sba, dsstox_cid_27882, dsstox_rid_82635, dsstox_gsid_47906, 1-butyl-2,3-dimethylimidazoliumchloride, 1-butyl-2,3-dime-thylimidazolium chloride
PubChem CID 2734167
IUPAC Name 1-butyl-2,3-dimethylimidazol-3-ium;chloride
SMILES CCCCN1C=C[N+](=C1C)C.[Cl-]

Specifications

Melting Point 89.5°C to 93.2°C
Quantity 50 g
Formula Weight 188.69
Percent Purity 99%
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium chloride
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only