accessibility menu, dialog, popup

UserName

CAS RN 98717-15-8

IUPAC Name: (2S)-N-(2,6-dimethylphenyl)-1-propylpiperidine-2-carboxamide hydrochloride
Synonyms: ropivacaine hydrochloride
Molecular Weight (g/mol): 310.87
Molecular Formula: C17H27ClN2O
InChi Key: NDNSIBYYUOEUSV-RSAXXLAASA-N
SMILES: Cl.CCCN1CCCC[C@H]1C(=O)NC1=C(C)C=CC=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)