accessibility menu, dialog, popup

UserName

CAS RN 98437-23-1

IUPAC Name: (1-benzothiophen-2-yl)boronic acid
Synonyms: 1-benzothiophen-2-ylboronic acid
Molecular Weight (g/mol): 178.01
Molecular Formula: C8H7BO2S
InChi Key: YNCYPMUJDDXIRH-UHFFFAOYSA-N
SMILES: OB(O)C1=CC2=CC=CC=C2S1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (4)
  • (1)
Physical Form 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (5)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)