missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-boronic acid, 97%

Catalog No. AAB2283514
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2283514 25 g
AAB2283503 1 g
AAB2283506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2283514 Supplier Thermo Scientific Chemicals Supplier No. B2283514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98437-23-1 | C8H7BO2S | 178.01 g/mol

Benzo[b]thiophene-2-boronic acid is used in the synthesis of phosphorescent sensor for quantification of copper(II) ion, UV promoted phenanthridine syntheses, Preparation of CYP11B1 inhibitors for treatment of cortisol dependent diseases, PDE4 inhibitors, Suzuki-Miyaura cross-coupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 98437-23-1
Molecular Formula C8H7BO2S
Molecular Weight (g/mol) 178.01
MDL Number MFCD01075674
InChI Key YNCYPMUJDDXIRH-UHFFFAOYSA-N
Synonym 2-benzothienylboronic acid, benzo b thiophen-2-ylboronic acid, benzo b thiophene-2-boronic acid, benzothiophene-2-boronic acid, 1-benzothien-2-ylboronic acid, thianaphthene-2-boronic acid, benzo b thiophene-2-ylboronic acid, 2-benzothiopheneboronic acid, benzothiophen-2-ylboronic acid
PubChem CID 2359
SMILES OB(O)C1=CC2=CC=CC=C2S1

Specifications

Melting Point ∼255°C
Quantity 25 g
Beilstein 1637924
Solubility Information Insoluble in water.
IUPAC Name 1-benzothiophen-2-ylboronic acid
Formula Weight 178.02
Percent Purity 98%
Chemical Name or Material Benzo[b]thiophene-2-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only