missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B28931G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28931G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28931G Supplier TCI America Supplier No. B28931G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98437-23-1
Molecular Formula C8H7BO2S
Molecular Weight (g/mol) 178.01
MDL Number MFCD01075674
InChI Key YNCYPMUJDDXIRH-UHFFFAOYSA-N
Synonym 2-benzothienylboronic acid, benzo b thiophen-2-ylboronic acid, benzo b thiophene-2-boronic acid, benzothiophene-2-boronic acid, 1-benzothien-2-ylboronic acid, thianaphthene-2-boronic acid, benzo b thiophene-2-ylboronic acid, 2-benzothiopheneboronic acid, benzothiophen-2-ylboronic acid
PubChem CID 2359
IUPAC Name (1-benzothiophen-2-yl)boronic acid
SMILES OB(O)C1=CC2=CC=CC=C2S1

Specifications

Melting Point 260°C
Color White-Yellow
Quantity 1 g
Formula Weight 178.01
Physical Form Crystalline Powder
Chemical Name or Material Benzo[b]thiophene-2-boronic Acid (contains varying amounts of Anhydride)