missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl chloride, 99%, pure, AcroSeal™

Catalog No. AC220271000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC220271000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC220271000 Supplier Thermo Scientific Chemicals Supplier No. 220271000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98-88-4 | C7H5ClO | 140.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-88-4
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
InChI Key PASDCCFISLVPSO-UHFFFAOYSA-N
Synonym benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
PubChem CID 7412
ChEBI CHEBI:82275
IUPAC Name benzoyl chloride
SMILES C1=CC=C(C=C1)C(=O)Cl

Specifications

Melting Point -1.0°C
Density 1.2100g/mL
Boiling Point 198.0°C
Flash Point 93°C
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.5520 to 1.5540
Linear Formula C6H5COCl
Quantity 100 mL
Beilstein 09,182
Fieser 01,50; 05,24; 13,219
Merck Index 15,1115
Specific Gravity 1.21
Solubility Information Solubility in water: reacts, 2 mg/mL. Other solubilities: decomposes in alcohols and in water, soluble in ether, benzene; miscible with oils,, carbon disulfide, ccl4
Viscosity 0.0012 Pas (30°C)
Formula Weight 140.57
Percent Purity 99%
Grade Pure
Chemical Name or Material Benzoyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 202-710-8

RUO – Research Use Only