CAS RN 98-47-5
IUPAC Name:
3-nitrobenzene-1-sulfonic acid
Synonyms:
3-nitrobenzenesulfonic acid
Molecular Weight (g/mol):
203.17
Molecular Formula:
C6H5NO5S
InChi Key:
ONMOULMPIIOVTQ-UHFFFAOYSA-N
SMILES:
OS(=O)(=O)C1=CC=CC(=C1)[N+]([O-])=O
1
–
1
of
1
results
1
3-Nitrobenzenesulfonic Acid Hydrate 98.0+%, TCI America™
CAS: 98-47-5 Molecular Formula: C6H5NO5S Molecular Weight (g/mol): 203.168 MDL Number: MFCD00064383 InChI Key: ONMOULMPIIOVTQ-UHFFFAOYSA-N Synonym: 3-nitrobenzenesulfonic acid,m-nitrobenzenesulfonic acid,benzenesulfonic acid, 3-nitro,3-nitrobenzenesulphonic acid,m-nitrobenzensulfonic acid,3-nitrophenylsulfonic acid,kyselina 3-nitrobenzensulfonova,benzenesulfonic acid, m-nitro,ccris 3131,kyselina nitrobenzen-m-sulfonova PubChem CID: 7387 SMILES: C1=CC(=CC(=C1)S(=O)(=O)O)[N+](=O)[O-]