missing translation for 'onlineSavingsMsg'
Learn More

3-Nitrobenzenesulfonic Acid Hydrate 98.0+%, TCI America™

Catalog No. N01385G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N01385G 5 g
N013825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N01385G Supplier TCI America Supplier No. N01385G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 98-47-5
Molecular Formula C6H5NO5S
Molecular Weight (g/mol) 203.168
MDL Number MFCD00064383
InChI Key ONMOULMPIIOVTQ-UHFFFAOYSA-N
Synonym 3-nitrobenzenesulfonic acid, m-nitrobenzenesulfonic acid, benzenesulfonic acid, 3-nitro, 3-nitrobenzenesulphonic acid, m-nitrobenzensulfonic acid, 3-nitrophenylsulfonic acid, kyselina 3-nitrobenzensulfonova, benzenesulfonic acid, m-nitro, ccris 3131, kyselina nitrobenzen-m-sulfonova
PubChem CID 7387
SMILES C1=CC(=CC(=C1)S(=O)(=O)O)[N+](=O)[O-]

Specifications

Color White-Yellow
Quantity 5 g
UN Number 2305
Formula Weight 203.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Nitrobenzenesulfonic Acid Hydrate
EINECSNumber (3)-2005
RTECSNumber DB7190000
TSCA Yes