accessibility menu, dialog, popup

UserName

CAS RN 98-05-5

IUPAC Name: phenylarsonic acid
Synonyms: phenylarsonic acid arsonic acid, as-phenyl- kyselina benzenarsonova PhAsO(OH)2 C6H7AsO3 monosodium salt benzenearsonic acid disodium salt benzenearsonic acid
Molecular Weight (g/mol): 202.04
Molecular Formula: C6H7AsO3
InChi Key: LVKZSFMYNWRPJX-UHFFFAOYSA-N
SMILES: O[As](O)(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)