missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsonic acid, 97%

Catalog No. AAA1062522
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1062522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1062522 Supplier Thermo Scientific Chemicals Supplier No. A1062522
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 98-05-5 | C6H7AsO3 | 202.041 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 98-05-5
Molecular Formula C6H7AsO3
Molecular Weight (g/mol) 202.041
MDL Number MFCD00002097
InChI Key LVKZSFMYNWRPJX-UHFFFAOYSA-N
Synonym benzenearsonic acid, monophenylarsonic acid, arsonic acid, phenyl, unii-57f9ku116m, kyselina benzenarsonova czech, arsonic acid, as-phenyl, kyselina benzenarsonova, benzenarsonic acid, benzenearsonie acid, acmc-20ajwq
PubChem CID 7365
ChEBI CHEBI:29851
IUPAC Name phenylarsonic acid
SMILES C1=CC=C(C=C1)[As](=O)(O)O

Specifications

Melting Point 158°C to 162°C (decomposition)
Quantity 100 g
UN Number UN3465
Merck Index 14,1067
Formula Weight 202.04
Percent Purity 97%
Chemical Name or Material Phenylarsonic acid
Hazard Category H301+H331
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P310-P304+P340-P311-P330-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: I
EINECSNumber 202-631-9
RTECSNumber CY3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only