accessibility menu, dialog, popup

UserName

CAS RN 98-03-3

IUPAC Name: thiophene-2-carbaldehyde
Synonyms: thiophene-2-carboxaldehyde
Molecular Weight (g/mol): 112.15
Molecular Formula: C5H4OS
InChi Key: CNUDBTRUORMMPA-UHFFFAOYSA-N
SMILES: O=CC1=CC=CS1
Boiling Point 
  • (3)
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (1)