accessibility menu, dialog, popup

UserName

H3CNH3CSSSSNCH3CH3

CAS RN 97-77-8

IUPAC Name: N,N-diethyl[(diethylcarbamothioyl)disulfanyl]carbothioamide
Synonyms: disulfiram tetraethylthiuram dicupral antietil exhoran espenal dupont fungicide 4472 esperal abstensyl (argentina) exhorran
Molecular Weight (g/mol): 296.52
Molecular Formula: C10H20N2S4
InChi Key: AUZONCFQVSMFAP-UHFFFAOYSA-N
SMILES: CCN(CC)C(=S)SSC(=S)N(CC)CC
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)