missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylthiuram disulfide, 97%

Catalog No. AAB2072130
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2072130 250 g
AAB207210B 1000 g
AAB207210I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2072130 Supplier Thermo Scientific Chemicals Supplier No. B2072130

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 97-77-8 | C10H20N2S4 | 296.524 g/mol

Dopamine beta-hydroxylase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-77-8
Molecular Formula C10H20N2S4
Molecular Weight (g/mol) 296.524
MDL Number MFCD00009048
InChI Key AUZONCFQVSMFAP-UHFFFAOYSA-N
Synonym disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
PubChem CID 3117
ChEBI CHEBI:4659
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC

Specifications

Melting Point 69°C to 72°C
Density 1.3
Boiling Point 117°C (17 mmHg)
Quantity 250 g
UN Number UN3077
Beilstein 1712560
Merck Index 14,3364
Formula Weight 296.54
Percent Purity 97%
Chemical Name or Material Tetraethylthiuram disulfide
Hazard Category H302-H317-H373
Hazard Statement GHS H Statement
H373-H302-H317
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P270-P272-P280g-P301+P312-P302+P352-P314-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 202-607-8
RTECSNumber JO1225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only