accessibility menu, dialog, popup

UserName

CAS RN 961-29-5

IUPAC Name: (2E)-1-(2,4-dihydroxyphenyl)-3-(4-hydroxyphenyl)prop-2-en-1-one
Synonyms: 2',4,4'-trihydroxychalcone 6'-deoxychalcone isoliquiritigenin trihydroxychalcone
Molecular Weight (g/mol): 256.26
Molecular Formula: C15H12O4
InChi Key: DXDRHHKMWQZJHT-FPYGCLRLSA-N
SMILES: OC1=CC=C(\C=C\C(=O)C2=CC=C(O)C=C2O)C=C1
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results